Agarose Type C

Supplier: MilliporeSigma

121852-100GM 121852-100GM
CA80057-770EA 1081.32 CAD
80057-770 CA80057-770
Agarose Type C
Agarose
MDL Number: MFCD00081294
CAS Number: 9012-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR