Adenine

Supplier: MilliporeSigma

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

1152-25GM 1152-100GM 1152-10GM
CA80057-762EA 286.33 CAD
CA80057-762 CA80108-034 CA80108-036
Adenine
Adenine
Formula: C₅H₅N₅
MW: 135.13 g/mol
MDL Number: MFCD00041790
CAS Number: 73-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR