4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)

Supplier: MilliporeSigma

Synonyms: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate

4876-1GM 4876-5GM
CA80057-148EA 121.28 CAD
CA80057-148 CA80057-150
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)
Formula: C₆H₄NNa₂O₆P·6H₂O
MDL Number: MFCD00066288
CAS Number: 333338-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR