n-Dodecyl-β-D-maltoside, ULTROL®

Supplier: MilliporeSigma

324355-1GM 324355-25GM 324355-5GM
CA80056-364EA 420.62 CAD
CA80056-364 CA80056-366 CA80056-370
n-Dodecyl-β-D-maltoside, ULTROL®
n-Dodecyl-β-D-maltoside
Formula: C₂₄H₄₆O₁₁
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR