n-Decyl-β-D-maltoside, ULTROL®

Supplier: MilliporeSigma

252718-1GM 252718-5GM
CA80056-270EA 572.55 CAD
CA80056-270 CA80056-272
n-Decyl-β-D-maltoside, ULTROL®
n-Decyl-β-D-maltoside
Formula: C₂₂H₄₂O₁₁
MDL Number: MFCD00061624
CAS Number: 82494-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR