Imidazole, ULTROL®

Supplier: MilliporeSigma

4015-25GM 4015-100GM
CA80055-294EA 468.49 CAD
CA80055-294 CA80055-566
Imidazole, ULTROL®
Imidazole
Formula: C₃H₄N₂
MDL Number: MFCD00005183
CAS Number: 288-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR