Digitonin, high purity

Supplier: MilliporeSigma

300410-1GM 300410-250MG 300410-5GM
CA80055-224EA 1225.99 CAD
CA80055-224 CA80055-226 CA80055-228
Digitonin, high purity
Digitonin
Formula: C₅₆H₉₂O₂₉
MDL Number: MFCD00077729
CAS Number: 11024-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR