Uridine-5′-diphosphogalactose disodium salt
Supplier: MilliporeSigma
Synonyms:
Udp-alpha-d-galactose disodium salt, UDP-galactose, Uridine-5'-diphospho-a-D-galactose disodium salt, UDP-Ga
670111-10MG
670111-50MG
CA80054-960EA
169.37
CAD
CA80054-960
CA80054-962
Uridine-5′-diphosphogalactose disodium salt
Uridine-5′-diphosphogalactose disodium salt
Formula:
C₁₅H₂₃N₂Na₂O₁₇P₂ MW: 611.27 g/mol |
MDL Number:
MFCD00077895 CAS Number: 137868-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...