Lecithin (from soy bean)

Supplier: MilliporeSigma

429415-100GM 429415-100GM
CA80052-866EA 930.79 CAD
CA80052-866 80052-866
Lecithin (from soy bean)
Lecithin
MDL Number: MFCD00147406
CAS Number: 8002-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR