Adenosine-5'-diphosphate monosodium salt

Supplier: MilliporeSigma

1171-1GM 1171-5GM
CA80050-104EA 366.29 CAD
CA80050-104 CA80050-106
Adenosine-5'-diphosphate monosodium salt
Adenosine-5'-diphosphate monosodium salt
Formula: C₁₀H₁₄N₅NaO₁₀P₂
MDL Number: MFCD00065987
CAS Number: 20398-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR