Nonidet® P 40 Substitute (NP-40) solution 10%, PROTEIN GRADE® detergent, sterile-filtered

Supplier: MilliporeSigma

492018-50ML 492018-500ML
CA16003-794EA 323.97 CAD
CA16003-794 CA16003-796
Nonidet® P 40 Substitute (NP-40) solution 10%, PROTEIN GRADE® detergent, sterile-filtered
Nonidet® P 40 Substitute (NP-40)
MDL Number: MFCD00132851
CAS Number: 9016-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR