Fluval® C3 Power Filter
Supplier: ROLF C. HAGEN (USA) CORP.
Fluval®
14015 14012 14002 14006 14024 14009
470347-650EA 32.5 CAD
470347-650 470347-630 470347-838 470347-816 470347-768 470347-658
Fluval® C3 Power Filter
Tanks Aquarium Tanks
Engineered for superior water quality.
  • Extensive 5-stage filter
  • Two mechanical, one chemical and two biological filter chambers
  • Slower water circulation resulting in cleaner aquarium water
  • Ideal for use in 75 to 190 L (20 to 50 US gal.) fresh or marine aquariums
Engineered to create superior water quality, the Fluval® C3 power filter provides extensive 5-stage filtration (two mechanical, one chemical and two biological stages) for outstanding results. The power filter is designed to provide maximum surface area and optimum dwell times in all stages of the filtration process.
Rigorous 2-stage mechanical filtration is performed by a thick layer of poly/foam that effectively traps large and fine debris. The foam slides out easily for quick cleaning. Activated carbon, located in an easily-manageable and removable basket, performs thorough chemical filtration and effectively removes toxins. In the final two stages, a Bio-Screen pad provides massive biological surface area for friendly bacteria to colonize, plus it blocks debris from entering a trickle chamber filled with C-Nodes that are super-charged for fast and efficient nitrification. The result is clean, clear and healthy water for aquarium life to thrive in.
The power filter also features a patented refiltration control system that lets you slow down the water output, creating a gentle flow to protect delicate fish and plants. At the same, a slower water circulation increases water dwell time in the filter, resulting in cleaner aquarium water. The Power Filter's completely modular design allows you to manage each stage separately and make changes at different times to maintain continuous biological activity. This results in a stress-free environment and ensures clear aquarium water.