Fluval® Water Testing Kits
Supplier: ROLF C. HAGEN (USA) CORP.
Fluval®
A7871 A7872 A7867 A7868 A7870 A7869
470347-690KT 20.4 CAD
470347-690 470347-716 470347-670 470347-858 470347-862 470347-832
Fluval® Water Testing Kits
Tanks Aquarium Tanks
Make sure your water is balanced.
  • Master kit or individual kits
The Fluval® Mini master test kit offers essential testing of pH, Ammonia, Nitrite, and Nitrate to help maintain a balanced and healthy environment for fish and plant life. Additional individual test kits allow testing for pH, Ammonia, Nitrate, Nitrites, and Phosphates.