SpectraMax® Mini Microplate Readers, Multi-Mode, Molecular Devices
Supplier: MOLECULAR DEVICES LLC. CA
5089096 5089097 5089098 5089099 5089100 5089102 5089103 5089104 5089334 SMAX-MINI-AF SMAX-MINI-AFL SMAX-MINI-AL
CA76640-486EA 3192.44 CAD
CA76640-486 CA76640-488 CA76640-490 CA76640-492 CA76640-494 CA76640-496 CA76640-498 CA76640-500 CA76640-502 CA76640-504 CA76640-506 CA76640-508
SpectraMax® Mini Microplate Readers, Multi-Mode, Molecular Devices
Microplate Readers
The SpectraMax® Mini multi-mode microplate reader offers a budget friendly solution that gives you exactly what you need when you need it. With three modes of detection for UV/Vis absorbance, fluorescence and luminescence, the SpectraMax® Mini reader is user-upgradeable - from two to three modes, providing researchers with a versatile instrument that does not compromise on sensitivity or performance.
  • Industry leading software - enjoy an extensive library of protocols, advanced reader control, and easy data management and analysis with our 'best in class'
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature range 4 °C above ambient to 45 °C
W×D×H 400×320×350 mm
Weight 18 kg