ROTAVISC Oils for Rotational Viscometers, IKA
Supplier: IKA Works
25000397 25000398 25000399 25000434 25000435 25000436 25000437 25000466 25000467 25000468 25000469
76499-098EA 271.81 CAD
76499-098 76499-100 76499-102 76499-104 76499-106 76499-108 76499-110 76499-112 76499-114 76499-116 76499-118
ROTAVISC Oils for Rotational Viscometers, IKA
Lubricants
These certified standard silicone oils can be used to check the measurement accuracy of ROTAVISC.
Delivery information: Supplied with certificate.