3-Mercapto-2-methyl-1-pentanol mixture of diasteroisomers ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M3274-1G M3274-5G
TCM3274-1GEA 64.78 CAD
TCM3274-1G TCM3274-5G
3-Mercapto-2-methyl-1-pentanol mixture of diasteroisomers ≥97.0% (by GC, titration analysis)
3-Mercapto-2-methyl-1-pentanol
Formula: C₆H₁₄OS
Boiling Pt: 46°C
MDL Number: MFCD08741458
CAS Number: 227456-27-1
EINECS: 434-900-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 % (total of isomer)
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR