AlphaTec® Neoprene Isolator Gloves, 55-300 Series, Ansell
Supplier: Ansell Healthcare
55300090 55300100
76413-622CS 3456.5 CAD
76413-622 76413-624
AlphaTec® Neoprene Isolator Gloves, 55-300 Series, Ansell
Gloves
Non-clean/non sterile neoprene RABS/Isolator gloves for use in 152 mm (6") ports.
  • Effective protection for medium duty chemical environments
  • Port size: 152 mm (6")
  • Exceptional chemical resistance against most acids, alcohols, oils, lubricants, and hydrocarbons
  • Medium thickness (20 mil/0.51 mm) increases dexterity, which improves the ability to manipulate objects
  • Unlined for effective cleaning
AlphaTec® Neoprene RABS and Isolator gloves deliver exceptional chemical resistance against most acids, alcohols, oils, lubricants, and hydrocarbons. They offer strong physical protection against abrasions and good protection against cuts and punctures.
Certifications: CE 0493, EN 374:2003, EN 388:2003, EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 388:2016, EN 420:2003 + A1:2009, Category III, UKCA 0321.
Packaging: 1 pair per black sealed plastic bag; 10 pairs per shipper carton.