Transystem™ Bacteriology Transport Swabs

Supplier: Copan Diagnostics
108CVI.US 110CVI.US
76640-388CS 934.62 CAD
76640-388 76640-390
Transystem™ Bacteriology Transport Swabs
Swabs Transport Swabs
Rayon swab bacterilogical specimen transport and collection kit.

  • No activation step required
  • Eliminate contamination risk with tamper-evident color change seal
  • Maintain traceability with printed lot number, expiration date, and product description on every swab package and tube
  • 5.25" swab shaft enables easy access to difficult sampling sites
  • Closure type: plastic round bottom Venturi hourglass tube
  • Media type: Amies Agar Gel Medium without charcoal
  • Validated sterility

COPAN Transystem™ collection devices are the perfect solution for veterinary professionals who need efficient and reliable sample collection and transport for clinical specimens. Transystem™ swabs are available with gel Amies media and come in multiple swab sizes and shaft materials to accommodate various applications and specific collection sites.

The Transystem™ long swab shafts and shatterproof plastic tube allow for easy access to difficult sampling sites. The tubes are designed to fit conveniently in regular test tube racks, allowing for easy transport and storage. The lot number, expiration date, and product description are clearly marked on every swab package and tube to ensure proper inventory rotation, product traceability, and use within the prescribed shelf life.

Simply uncap, collect, and recap the tube. The swabs are pre-moistened and there are no glass ampules or seals to break, making the product simple, safe, and easy to use. The Venturi Hourglass tube design enhances performance and eliminates undesirable bubbles, breaking in the deep agar gel column, ensuring optimal specimen collection. Ensure media integrity with VI-PACK packaging made from high-tech composite plastic films that retards penetration of atmospheric oxygen preventing dehydration of liquid or agar gel medium. 100% plastic on plastic film eliminates fiber shed, important in surgical settings. All media have been extensively validated and tested for a low bioburden and contain less than two non-viable bacterial cells per 10 high power microscopic fields.

Certifications: Certificates of sterility and quality assurance are available upon request from COPAN.

Ordering information: Refrigerate at 2 to 8 °C or store at room temperature (20 to 25 °C) and process within 48 hours (except for Neisseria gonorrhoeae cultures which should be processed within 24 hours).

Packaging: 50 kits per pack, 500 kits per case, in recyclable VI-PACK packaging.

Caution: The product should be stored in its original container at 5 to 25 °C until used.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR