CELLSTAR® Cell-Repellent Surface Dishes, Greiner Bio-One
Supplier: Greiner Bio-One
664970 627979 628979
30618-024CS 116.27 CAD
30618-024 10014-312 10014-314
CELLSTAR® Cell-Repellent Surface Dishes, Greiner Bio-One
Culture Dishes Cell Culture Dishes
These sterile, PS cell culture dishes have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modification, the surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
  • Effectively inhibits cell adhesion
  • Sterile
  • Lid with condensation rings
  • Other formats available (flasks, microplates, multiwell plates)
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as spheroid cultures and stem cell aggregate formation, suspension culture of semi-adherent and adherent cell (eg. for suspension culture of macrophages), gel-based 3D cultures, or magnetic 3D cell culturing (Nano3D technology).