SatPax® Polx® Nonwoven Pre-wetted 100% Polyester Cleanroom Wipes, Berkshire
Supplier: Berkshire
SPXPNW00112 SSPPNW00124
76638-272CS 454.04 CAD
76638-272 76638-260
SatPax® Polx® Nonwoven Pre-wetted 100% Polyester Cleanroom Wipes, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® Polx® Nonwoven is a 100% polyester hydroentangled nonwoven wiper recommended for ISO Class 5 and above environments. It is an extremely soft and pliable wiper ideal for all types of sensitive surfaces and an excellent choice for wrapping delicate parts for protection.
  • Exceptional cleanliness
  • Prevents abrasion with its extremely soft and pliable surface
  • Increases cleaning protocol consistency
  • Lot to lot traceability
  • Reduces alcohol usage and preparation/handling costs
  • 70% IPA, 30% DI water, 60% saturation
  • Available in nonsterile and sterile formats
SatPax® Polx® Nonwoven is a 100% polyester hydroentangled nonwoven wiper recommended for ISO Class 5 and above environments. It is an extremely soft and pliable wiper ideal for all types of sensitive surfaces and an excellent choice for wrapping delicate parts for protection. This pre-wetted format provides a cost-effective and easy-to-use solution versus traditional bulk handling of solvents, maintenance of squirt bottles and inconsistent wetting and cleaning associated with wetting a dry wiper. Additionally, it's critically low fiber and particle generation and minimal non-volatile residues make SatPax® Polx® nonwoven an exceptional choice for streak-free cleaning.
Certifications: ISO Class 5 – 8. Cleanroom Class 100 – 100,000. EU Grade B/C/D. Sterile wipes are gamma irradiated and sterile validated to a 10¯⁶ Sterility Assurance Level per AAMI guidelines.
Ordering information: Certificate of Sterility with each case and available on the web by lot number 24/7. Details: Expiration Date, Radiation Dosage, Lot Information, Meets cGMP requirements for traceability