SatPax® MicroSeal®-VP Pre-wetted Sealed Edge Cleanroom Laundered 100% Polyester Knit Wipes, Berkshire
Supplier: Berkshire
SatPax® MicroSeal®-VP
SPXMSVP0028 SPXMSVP0068
76638-258CS 1220.23 CAD
76638-258 76638-262
SatPax® MicroSeal®-VP Pre-wetted Sealed Edge Cleanroom Laundered 100% Polyester Knit Wipes, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® MicroSeal®-VP is an ultrasonically sealed edge cleanroom laundered wiper recommended for ISO Class 3 and above environments.
  • Critically low particles, fibers, ions and extractables
  • High abrasion resistance
  • High Absorbency
  • Lightweight material reduces actual cost in use compared to heavier weight products
  • Increases cleaning protocol consistency
SatPax® MicroSeal®-VP is an ultrasonically sealed edge cleanroom laundered wiper composed of 100% continuous filament polyester knit fabric. The wiper is pre-wetted with a consistent concentration of IPA and DI water and available in a 100% IPA option. This combination of properties provides SatPax® MicroSeal ®-VP with the highest cleanliness, abrasion resistance, and chemical compatibility required for cleaning critical environments. The pre-wetted format offers a cost-effective and easy-to-use solution versus traditional bulk handling of solvents and maintenance of squirt bottles.
Certifications: ISO Class 3 – 5. Cleanroom Class 1 – 100. EU Grade B/C/D.