Polx® Nonwoven 100% Polyester Cleanroom Wiper, Berkshire
Supplier: Berkshire
PNW040440 PNW090920 PNW121220
76637-680CS 327.06 CAD
76637-680 76637-682 76637-678
Polx® Nonwoven 100% Polyester Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polx® Nonwoven is a 100% polyester hydroentangled nonwoven wiper recommended for ISO Class 5 and above environments.
  • 100% polyester hydroentangled nonwoven treated with proprietary process
  • No cellulose components, 100% synthetic
  • Chemically compatible with commonly used disinfectants and sporicides
  • Knife cut
  • Lot to lot traceability
  • Autoclavable
These are extremely soft and pliable wipers ideal for all types of sensitive surfaces, and an excellent choice for wrapping delicate parts for protection.
Certifications: ISO Class 5 to 8, Cleanroom Class 100More Product Information