Eisco® Slotted Masses, Zinc Casted
Supplier: Wards
PHZNW100 PHZNW010 PHZNW050 PHZNW020
470343-980EA 9.49 CAD
470343-980 470345-850 470346-098 470346-112
Eisco® Slotted Masses, Zinc Casted
Weights Calibration Weights
Individual Slotted Masses for placement on hangers are made of powder-coated, die-cast zinc and stamped with mass value for easy identification.
  • Masses die-cast in zinc and powder-coated to ensure durability and long life
  • Slotted for stacking on mass hangers (available separately)
  • Each piece is stamped with mass value for easy identification
  • Accuracy of ±2% make them an economical alternative to brass masses
  • Ideal as additional or spare part for slotted mass sets
Individual Slotted Masses are made of cast zinc and powder-coated to ensure durabilityMore Product Information