Eisco® Hooked Weights, Iron
Supplier: Wards
HKW10G HKW100 HKW20G HKW200 HKW50G HKW500
470343-914EA 7.9 CAD
470343-914 470344-262 470344-364 470344-386 470345-724 470345-752
Eisco® Hooked Weights, Iron
Weights Calibration Weights
Hooked Iron Weights with cylindrical bodies stamped with weight, cone-shaped tops, and bottom bar and slot to hang additional weights.
  • Cylindrical with cone-shaped top
  • Integral hook for easy lifting and hanging
  • Slotted bottom with a bar for hanging additional hooked weights underneath (excluding 10 g weight)
  • Each piece is stamped with weight for easy identification
  • Ideal as additional or spare part for hooked weight sets
Single hooked weights made of high-quality iron. Cylindrical with cone-shaped top. Features hook for easy lifting and hanging. Bottom slot allows for other hooked weights to be attached in tandem. Stamped with weight for easy identification.