Eisco® Balance Weights Set, Stainless Steel
Supplier: Wards
PH0037CSS PH0037BSS PH0037ASS
470344-394EA 236.35 CAD
470344-394 470344-450 470344-876
Eisco® Balance Weights Set, Stainless Steel
Weights Calibration Weights
Sets of stainless steel balance weights individually stamped and packed in a premium wooden storage block.
  • Individually calibrated balance weights in metric grams
  • Made from high-quality stainless steel
  • Features easy-to-handle, contoured shape
  • Each piece is stamped with weight for easy identification
  • Supplied in a premium wooden storage block
Sets include the most commonly used balance weights. Each is made from high-grade stainless steel and stamped with its weight. All sets include a polished wooden storage block with a place for each weight.