Celestron Lens Cleaning Kit
Supplier: CELESTRON INTERNATIONAL INC SE
Celestron
93576
470343-296EA 39.4 CAD
470343-296
Celestron Lens Cleaning Kit
Cleaning Agents Lens Cleaning Solutions
Wipe away dust, dirt, fingerprints, and other particles on lenses to see clearly with the Celestron Lens Cleaning Kit.
  • Provides streak-free results
  • Kit contains 10 lens wipes, lens cleaning fluid, a lens cloth, and a retractable lens brush
  • Easily transportable in a soft cloth pouch with a convenient belt loop
This cleaning kit works on all glass and optical surfaces, including telescopes, binoculars, spotting scopes, monoculars, microscopes, and even camera lenses, computer screens, and mobile devices. The kit comes complete with a retractable lens brush to remove dust particles and dirt and two options to clean smudges and fingerprints off of optics, pre-moistened lens wipes, and a bottled lens cleaning fluid with soft cloth.
NOTE: The lens cleaning fluid, cloth, and wipes can be used on corrector lens of Schmidt-Cassegrain telescopes. These items should NOT be used on the primary or secondary mirrors of telescopes.
The durable carrying pouch is divided into three sections to keep all the items organized and close at hand. The Lens Cleaning Kit can go anywhere with the convenient belt loop. It also fits perfectly in a pocket or a small bag. Add it to an adventure kit today and enjoy better views.
Delivery information: Kit includes 10 lens wipes (individually wrapped), lens cleaning fluid (30 ml), a lens cloth, and a retractable lens brush.