Bacto™ Proteose Peptone No. 2, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
212110 212120
CA76628-896EA 11406.91 CAD
CA76628-896 CA76628-736
Bacto™ Proteose Peptone No. 2, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Bacto Proteose Peptone No. 2 is member of the Bacto Proteose Peptones, which are enzymatic digests of protein.
  • Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
  • Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
It offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of Fastidious microorganisms
Studies of peptic digests of animal tissue prepared under varying digestion parameters demonstrated that no single peptone is the most suitable nitrogen source for every microbiological application. These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone.
We also offer alternatives to Bacto Proteose Peptones for scale-up to production applications, including BiTek Proteose Peptone.
Certifications: LUO