Bacto™ Casamino Acids, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
223020 223030
CA76628-904EA 1239.1 CAD
CA76628-904 CA76628-944 CA76628-910
Bacto™ Casamino Acids, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto Casamino Acids is an animal origin (AO) acid hydrolysis of casein. This supplement contains low salt and iron content making it an excellent supplement for media formulations where nitrogen requirements are minimal.
  • An animal origin (AO) acid hydrolysis of casein
  • Contains low salt and iron content, making it an excellent supplement for media formulations where nitrogen requirements are minimal
  • Maintains many of the nutritional benefits of hydrolyzed casein, but is further supplemented with growth factors, cystine, maltose, and an optimum amount of iron and inorganic salts
  • Supports the growth of Bordetella, Corynebacterium, Pseudomonas, Haemophilus, and Escherichia species
  • Successfully used in fermentation applications for Pneumococcal, Diptheria, Pertussis, Tetanus and Zoster vaccines
  • Together with Bacto Yeast Extract, technical, this supplement has been successfully used in the production of animal and human health vaccines
It maintains many of the nutritional benefits of hydrolyzed casein, but is further supplemented with growth factors, cystine, maltose, and an optimum amount of iron and inorganic salts.