Resurge™ CD5 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
670024 670026
CA76628-878EA 359.63 CAD
CA76628-878 CA76628-734 CA76628-834
Resurge™ CD5 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Resurge CD5 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media.
  • Used in CHO mammalian culture systems
  • Can be used either as a media supplement or a culture system feed
This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Resurge CD5 Supplement, without glucose and L-glutamine, was developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.