Resurge™ CD4 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Resurge CD4 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media.
- Used in CHO mammalian culture systems
- Can be used either as a media supplement or a culture system feed
This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Resurge CD4 Supplement, without glucose and L-glutamine, was developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.