Resurge™ CD3 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
670020
CA76628-816EA 251.67 CAD
CA76628-816 CA76628-884 CA76628-940
Resurge™ CD3 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Resurge CD3 Supplement, without glucose and L-glutamine, is an animal origin-free (AOF) protein-free chemically defined supplement. It can be used in batch and fed batch mode in cell culture processes.
  • Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications
  • Can be used in batch and fed batch mode in cell culture processes
  • Use in bacterial applications
Resurge CD3 Supplement is formulated without glucose, L-glutamine, and sodium bicarbonate. Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications. All components are chemically defined with known molecular formulae and structures and meet our stringent Animal Origin-Free (AOF) specification.