Resurge™ CD1 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
670013
CA76628-778EA 319.98 CAD
CA76628-778 CA76628-856 CA76628-948
Resurge™ CD1 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Resurge CD1 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media.
  • Used in CHO mammalian culture systems
  • Can be used either as a media supplement or a culture system feed
This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Resurge CD1 Supplement, without glucose and L-glutamine, was developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.