Recharge™ Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
670003
CA76628-814EA 182.88 CAD
CA76628-814 CA76628-746 CA76628-784
Recharge™ Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Recharge Supplement, without glucose and L-glutamine, is a chemically defined, animal origin-free (AOF) protein-free supplement designed to increase protein yields, while maintaining acceptable viability and cell growth.
  • A chemically defined (CD) supplement to increase protein yields, while maintaining acceptable viability and cell growth
  • Developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors
  • Use in CHO mammalian cell culture systems, as a media supplement or a culture system feed
  • Has been beneficial for non-CHO mammalian systems and microbial systems
  • Can be used as a replacement for yeast extract
The supplement has been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations. Thereforce, these two components should be supplements by the user and monitored as appropriate.