Polypeptone™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
211910
CA76628-916EA 161.24 CAD
CA76628-916 CA76628-906
Polypeptone™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Polypeptone Peptone is a mixture of peptones made up of equal parts of pancreatic digest of casein and peptic digest of animal tissue.
  • Animal origin mixture of peptones
  • Includes the high content of amino acids
Found to meet nutritional requirements of various bacteria, fungi, and mammalian cells for which a single source of casein or meat peptones has been unsatisfactory.
Polypeptone Peptone includes the high content of amino acids and small polypeptides characteristic of pancreatic digest of casein and the larger polypeptides characteristic of peptic digest of animal tissue.