OneFeed™ Supplement, Gibco™ (a part of Thermo Fisher Scientific)
CA76628-872EA 2156.34 CAD
CA76628-872 CA76628-846 CA76628-798
OneFeed™ Supplement, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Gibco™ OneFeed™ Supplement is a versatile chemically defined, animal origin–free, protein-free feed that can be used with a variety of media to enhance protein.
  • Chemically defined (CD) and animal origin-free (AOF)
  • Developed using several different cell lines in various culture systems with a variety of commercially available media
  • Balanced nutritional profile, providing key components to support high growth and production throughout the culture life
  • Can be used a feed supplement to enhance growth and production of a variety of CHO lines
This high-performance feed has been developed using several different cell lines in various culture systems with a variety of commercially available media. OneFeed Supplement has a balanced nutritional profile that provides key components to support high growth and production throughout the culture life.