Difco™ TC Yeastolate, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
670079 292804
CA76628-770EA 39639.86 CAD
CA76628-770 CA76628-750 CA76628-836
Difco™ TC Yeastolate, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Difco TC Yeastolate, ultra-filtered (UF), is an animal origin-free (AOF) product derived from water-soluble portions of autolyzed yeast (Saccharomyces cerevisiae). This product is intended as a nutritional supplement for bacterial, insect, and mammalian cell culture.
  • Mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), as well as vitamins
  • Meets additional requirements for reduced endotoxin, clarity, and suitability for the biopharmaceutical industry
  • Ultra-filtered
  • Supplementation of TC yeastolate to CHO cell culture can enhance growth and mAb production
These products are intended as nutritional supplements for bacterial, insect, and mammalian cell culture. TC yeastolate has been used in insect cell culture. TC yeastolate was found to be a very versatile supplement to enhance growth and production characteristics of Sf9 and Gibco™ High Five™ cells [1-5]. Additionally, supplementation of TC yeastolate to CHO cell culture can enhance growth and mAb production.