Bacto™ Soytone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
243610 243620
CA76628-738EA 2927.42 CAD
CA76628-738 CA76628-914
Bacto™ Soytone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto Soytone is an enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion. The naturally high concentrations of vitamins and carbohydrates found in soy make Bacto Soytone a nutritious and excellent carbohydrate source for those not concerned about the processing step involving an animal-based material.
  • An enzymatic digest of soy that uses an animal origin (AO)(porcine) enzyme for digestion
  • Naturally high concentrations of vitamins and carbohydrates
  • Valuable alternative for supplementing both mammalian and microbial processes
  • Effective in the recovery of stressed Escherichia coli
This supplement offers a unique nutritional profile as compared to other soy products, and so can be a valuable alternative to consider for supplementing both mammalian and microbial processes.
Certifications: LUO