Bacto™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
211692
CA76628-928EA 41270.05 CAD
CA76628-928 CA76628-922 CA76628-782 CA76628-858
Bacto™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein.
  • Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
  • Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
Bacto Proteose Peptone No. 4 offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.
Certifications: LUO, MUO.