VWR® Cleanroom Gloves Class 10 Nitrile Tacky Gloves
Supplier: VWR International
76533-206 76533-208 76533-210 76533-212 76533-214 76533-216
76533-206CS 382.99 CAD
76533-206 76533-208 76533-210 76533-212 76533-214 76533-216
VWR® Cleanroom Gloves Class 10 Nitrile Tacky Gloves
Gloves
VWR® Cleanroom nitrile gloves are engineered for superior protection and comfort in controlled environments. These tacky gloves meet stringent Class 10 (ISO 4) standards, ensuring minimal particulate contamination. Ideal for pharmaceutical, biotech, and electronics industries, they offer excellent chemical resistance and tactile sensitivity for critical cleanroom tasks.
  • Designed for elbow length protection
  • Ambidextrous
  • Micro textured fingers and palm, beaded cuff
  • Not made from natural rubber latex
  • Static-dissipative
  • Single-use only, ISO-4 compliant