VWR® Urine Collection Containers with Needle, 100 ml
Supplier: VWR International
76620-614 76620-616
76620-614CS 520.29 CAD
76620-614 76620-616
VWR® Urine Collection Containers with Needle, 100 ml
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
This urine collection containers with needle will keep a clean and secure sample transfer directly into urine tubes.
  • Closed system allows sample material to be transported, analyzed and stored without risk of contamination or leakage
  • Suitable for application of evacuated blood collection tubes with an external diameter less than 18 mm
  • Leak-resistant container features tight seal to help avoid spillage
The container features molded patient ID label and molded graduations in both ounces and milliliters.
Certifications: Manufactured under EN ISO 13485: 2016.