Multidrop® Combi Microplate Dispensers, Thermo Scientific
Supplier: Thermo Fisher Scientific
5840330 5840340
CA76638-948EA 33453.2 CAD
CA76638-948 CA76638-950
Multidrop® Combi Microplate Dispensers, Thermo Scientific
Microplate Dispensers
Based on the proven and reliable Multidrop™ range, the Multidrop™ Combi provides high speed bulk reagent dispensing for biotech and pharmaceutical laboratories. The Multidrop™ Combi is remarkably flexible, dispensing into plates ranging from 6-well to 1536-wells with automated dispensing height adjustment for low profile to deep well plates.
  • Precise dispensing with peristaltic pump technology across a wide range: 0.5 to 2500 µl
  • Reduced reagent costs with low volume dispensing and minimal dead volumes
  • Simple operation with icon-based graphic display
  • High
More Product Information