Eisco High-Performance Variable Volume Micropipettes
Supplier: Wards
VVM7 VVM5 VVM13 VVM2 VVM3 VVM1 VVM10 VVM4 VVM12 VVM9 VVM11 VVM8 VVM6
470343-626EA 228.65 CAD
470343-626 470343-726 470343-810 470343-816 470344-014 470344-184 470344-602 470344-854 470345-100 470345-366 470345-692 470345-790 470345-910
Eisco High-Performance Variable Volume Micropipettes
Pipettes Micropipettes
Variable volume micropipettes are fully autoclavable and feature a simple ergonomic design for comfort, color coded plungers, and calibration conforming to DIN ISO standards.
  • Soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use
  • Fully autoclavable with a durable, autoclavable PVDF tip cone
  • Slip-resistant thermoplastic elastomer (TPE) body grip to minimize body heat transfer to the micropipette during long use, resulting in higher accuracy
  • Color coded plungers for easy identification
  • Calibration conforms to DIN 12650 & EN-ISO 8655 standard ensuring high accuracy and precision - Calibration report enclosed with every micropipette
High performance variable volume micropipettes have a simple design which allows for easy adjustment and reading of increments.