Eppendorf® 5427 R Hydrocarbon Cooling Refrigerated Microcentrifuges
Supplier: Eppendorf
5429000010 5429000133 5429000273 5429000373
CA76637-330EA 12749.86 CAD
CA76637-330 CA76637-332 CA76637-334 CA76637-336
Eppendorf® 5427 R Hydrocarbon Cooling Refrigerated Microcentrifuges
Centrifuges Microcentrifuges
The refrigerated Eppendorf Centrifuge 5427 R supports your greener lab with hydrocarbon cooling. It's an ideal solution for laboratories with a variety of molecular biology applications or who are in need of high-throughput solutions for 1.5 to 2 ml tubes - all combined in a microcentrifuge with a high throughput/footprint-ratio.
  • Cooled with natural refrigerant (R290) to protect samples and the planet
  • Soft touch one-finger closure for ergonomic operation
  • FastTemp function for quick pre-cooling of the centrifuge, e.g., from ~23 to 4 °C in only 11 minutes
  • ECO shut-off can be programmed to engage after 1, 2, 4 or 8 hours of non-use for maximum energy savings and extended compressor life
  • Built-in condensation drain to eliminate water accumulation
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • No.of.rotors available: Nine rotors
The refrigerated microcentrifuge Centrifuge 5427 R was designed with high-throughput research applications in mind. With its aerosol-tight 48-place rotor it handles even the toughest challenges in a safe and reliable manner to provide the efficiency boost you need. With speeds of up to 25,001×g, this microcentrifuge is fast enough for all your molecular biology applications. All aerosol-tight rotors are equipped with the Eppendorf QuickLock lids for easy rotor lid opening and closing to enable safe centrifugation of hazardous samples. Choose the Centrifuge 5427 R if a 24-place microcentrifuge does not meet your needs regarding tube capacity and if you don't need the extra versatility our cross-over Centrifuge 5430R can offer.
Certifications: ACT Label Certificate, UL Listed, Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).