NPS® Wood Science Lab Tables, Phenolic Top, Book Compartments, National Public Seating
Supplier: National Public Seating
SLT1-2472PB SLT2-2472PB
76276-826EA 2645.14 CAD
76276-826 76276-838
NPS® Wood Science Lab Tables, Phenolic Top, Book Compartments, National Public Seating
Furniture Tables and Workbenches
NPS wood frame science table is an asset to any lab or classroom. The 24×72" phenolic top can withstand various chemicals. Rubber footing and adjustable glides protect floors and allow for accurate leveling. Book compartments are an added feature for storage.
  • Solid ash legs and apron
  • Legs attach with metal on metal double bolt through assembly on 13-gauge steel corner braces
  • Adjustable glides for accurate leveling
  • Black, ³/₄" thick phenolic top is highly impervious to most chemicals, acids, corrosions, stains and moisture
  • Comparable to epoxy resin tops
  • 10-year warranty
Phenolic science lab tables with book compartments will have perfect chemistry with any science classroom. These paragons of endurance are built with strong ash frame and side panel aprons beneath a solid, black, phenolic resin tabletop with a matte finish. The work surface is crack-resistant and can be modified using carbide-tipped tools. It is impervious to extreme cold, heat up to 275 °F as well as damage from virtually any chemical, acid or possibly corrosive substance that may be found in a laboratory setting. Includes two storage compartments concealable with the included side-panel aprons. Robust legs are spot-welded to the frame with metal-on-metal double-bolts on 13-gauge steel corner braces for maximal stability. The PSLT's feet are outfitted with rubber liquid-protective boots and adjustable glides for accurate floor leveling. This product meets or exceeds SEFA environmental industrial standards.
Book compartments measure 14¹/₄L×12³/₄D×4"H.
Certifications: ANSI-BIFMA Certified, MAS Certified Green.