Trinocular Stereo Microscopes with Moticam, Camera Bundle, SMZ-171-TLED
Supplier: Motic
1100501300053 1100501300051 1100501300052 1100501300051 1100501300052
CA76635-312EA 4021.8 CAD
77575-314 76635-310 CA76635-312 CA76635-310 76635-312
Trinocular Stereo Microscopes with Moticam, Camera Bundle, SMZ-171-TLED
Microscopes
With the new SMZ-171 Stereo Zoom microscope Motic proudly introduces an optically improved addition to its well-established SMZ Stereo series. New materials for ESD compatibility as well as optimized LED illumination options have been added to this series to create a versatile Stereo microscope platform for a wide range of biological and material science applications.
  • SMZ-171-TLED
  • Greenough zoom optical system
  • 45° Trinocular head, 360° rotation
  • Photo/video 50/50 fixed
  • Widefield adjustable eyepieces 10×/F.N. 23
  • 6.7:1 zoom rRatio, WD = 110 mm
  • Magnification range 0.75 to 5×
  • Moticam BMH4000X
  • 1920×1080 HDMI monitor with attachable microscopy camera with 4K (3840×2160) resolution in SD card
  • 4K through HDMI
  • 20 fps at 3840×2160 through USB connection
  • 12 V power adapter, CS ring, Motic 4-dot calibration slide, dust cap, manual, 12 mm focusable lens, 30 mm and 38 mm eyepiece adapters, macro tube, macro calibration card
  • Motic Images Plus 3.1 for Windows/macOS
  • Moticam BTI10
  • 10" LCD Android tablet with 1280×800 screen resolution
  • Connection Wi-Fi (2.4 and 5 GHz), Ethernet
  • WiFi Resolution 4 MP (2592×1520 pixels) captured resolution
  • WiFi output (802.1 a/b/g/n/ac supporting both hot-spot mode or using existing WiFi network)
  • Preloaded optimized MotiConnect app
The Moticam BMH4000X is a camera that was designed for ease of use and the ability to output fast live images at 4K (3840×2160) pixels in full HD. Unlike other cameras, the Moticam BMH4000X can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through the built-in 11.6" LCD screen, or connect your camera through USB to a Windows, Mac or Linux computer.
The Moticam BTI10 unlike other Moticams is combined with a 4.0 MP and a 10" Android tablet that will deliver fast live images up to 30 fps on any trinocular microscope with the right C-mount adapter. With the convenience of a detachable tablet and WiFi image data transfer, the flexibility to capture images and bring your images anywhere has never been so easy.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Delivery information: Digital camera bundle comes with SMZ-171-TLED Trinocular, Moticam BMH4000 or Moticam BTI10, compatible C-mount adapter.