Trinocular Compound Microscopes with Moticam, Camera Bundle, Panthera C2
Supplier: Motic
1100403300222 1100403300221 1100403300223 1100403300221 1100403300222
CA76635-308EA 4585.8 CAD
CA76635-308 77575-312 CA76635-306 76635-308 76635-306
Trinocular Compound Microscopes with Moticam, Camera Bundle, Panthera C2
Microscopes
If it is about stained sections with cover glass protection, about native smears, fine needle biopsies, blood smears which wanted to be treated with or without cover glass: Panthera C2 with its selection of specified optics is your solution. The new Ultra Contrast (UC) objectives with additional glass elements to reach a 22 mm Field of View, working perfectly together with neutral eyepieces (thanks to CCIS© a fully corrected intermediate image for digital access is granted) guarantee best performance in terms of color fidelity and contrast. Microscope hardware, illumination, handling: Simply convincing.
  • Panthera C2
  • Trinocular head Siedentopf type 25° inclined, 360° swiveling
  • UC Widefield high eyepoint eyepiece UC-WF10×/22 mm
  • With diopter adjustment on both eyepieces and rubber eyecups
  • Reversed coded quintuple nosepiece with light tracer (intelli Light Management)
  • LED light indicator
  • Moticam BMH4000X
  • 1920×1080 HDMI monitor with attachable microscopy camera with 4K(3840×2160) resolution in SD Card
  • 4K through HDMI
  • 20 fps at 3840×2160 through USB connection
  • 12 V power adapter, CS ring, Motic 4-dot calibration slide, dust cap, manual, 12 mm focusable lens, 30 and 38 mm eyepiece adapters, macro tube, macro calibration card
  • Motic Images Plus 3.1 for Windows/macOS
  • Moticam BTI10
  • 10" LCD Android tablet with 1280×800 screen resolution
  • Connection Wi-Fi (2.4 and 5 GHz), Ethernet
  • WiFi resolution 4 MP (2592×1520 pixels) captured resolution
  • WiFi output (802.1 a/b/g/n/ac supporting both hot-spot mode or using existing WiFi network)
  • Preloaded optimized MotiConnect app
The Moticam BMH4000X is a camera that was designed for ease of use and the ability to output fast live images at 4K (3840×2160) pixels in full HD. Unlike other cameras, the Moticam BMH4000X can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through the built-in 11.6" LCD screen, or connect your camera through USB to a Windows, Mac or Linux computer.
The Moticam BTI10 unlike other Moticams is combined with a 4.0 MP and a 10" Android tablet that will deliver fast live images up to 30 fps on any trinocular microscope with the right C-mount adapter. With the convenience of a detachable tablet and WiFi image data transfer, the flexibility to capture images and bring your images anywhere has never been so easy.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Delivery information: Digital camera bundle comes with Panthera C2 Trinocular, Moticam BMH4000 or Moticam BTI10, compatible C-mount adapter.