Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Supplier: VALUE PLASTICS TE
SFMN655-6005100 SFMN670-6005100 SFMN680-6005100 MFLX31515-93BLK SFMX655-6005100 SFMX6110-6005250 SFMX670-6005100 SFMN6100-6005100 SFMX680-600550 SFMXC-6005
MFLX31515-95BLKEA 1415.68 CAD
MFLX31515-95BLK MFLX31515-89BLK MFLX31515-90BLK MFLX31515-93BLK MFLX31515-97BLK MFLX31515-99BLK MFLX31515-91BLK MFLX31515-96BLK MFLX31515-92BLK MFLX31513-18BLK
Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Fittings and Connectors
Certificate of compliance included!
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilizable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow—and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemicalMore Product Information