Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate
Supplier: VALUE PLASTICS TE
MTLL210-91000 MTLL220-91000 MTLL230-91000 MTLL240-91000 MTLL410-91000 MTLL250-91000 MTLL055-91000 MLRL055-91000
MFLX45504-00BLKEA 571.96 CAD
MFLX45504-00BLK MFLX45504-02BLK MFLX45504-04BLK MFLX45504-06BLK MFLX45504-07BLK MFLX45504-08BLK MFLX45504-19BLK MFLX45504-50BLK
Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate
Fittings and Connectors
Ensure your process integrity with full material traceability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly
More Product Information