Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Nylon
Supplier: VALUE PLASTICS TE
MTLL210-11000 MTLL220-11000 MTLL230-11000 MTLL240-11000 MTLL250-11000 MTLL055-11000 MTLS210-11000 MTLL004-11000 MTLL007-11000 MTLL013-11000
MFLX45505-00BLKEA 472.42 CAD
MFLX45505-00BLK MFLX45505-02BLK MFLX45505-04BLK MFLX45505-06BLK MFLX45505-08BLK MFLX45505-19BLK MFLX45505-22BLK MFLX45505-31BLK MFLX45505-32BLK MFLX45505-33BLK
Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Nylon
Fittings and Connectors
Ensure your process integrity with full material traceability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.