Value Plastics® Adapter Fittings, Luer to Plug
Supplier: VWR International
MFLX45518-70BLK MTLLP-60051000 MTLSP-60051000
MFLX45518-70BLKEA 496.12 CAD
MFLX45518-70BLK MFLX30800-30BLK MFLX45518-58BLK
Value Plastics® Adapter Fittings, Luer to Plug
Fittings and Connectors
Full material traceability to ensure process integrity.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.